Assay Details
| Gene Symbol: Ubap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111145.1 | NP_001104615.1 | 6 - 7 | 1247 | 74 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032493 | NM_001111145.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267056 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555855 | AK144887.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555855 | AK168948.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555855 | BC011501.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555855 | BC062103.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555855 | NM_145916.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19004 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032493 | NM_001111145.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07587209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07470671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07457509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07746534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07523590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07523589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07587217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08088680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07457513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08192014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07587223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07457508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08123997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07527745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07470670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07587210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07934012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07587213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Murine Genome U74v2 Set | 138145_PM_at | - | - |
| Murine Genome U74v2 Set | 138145_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331199 | - | - |

Ubap1l
Genomic Map