Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290528.1 | NP_001277457.1 | 5 - 6 | 1991 | 60 | |
| GenBank mRNA | BC046514.1 | - | 2 - 3 | 387 | 60 | |
| BC057197.1 | - | 4 - 5 | 700 | 60 | ||
| BC078445.1 | - | 5 - 6 | 1030 | 60 | ||
More Information
Set Membership: |
|

Map3k10
Genomic Map