Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290528.1 | NP_001277457.1 | 8 - 9 | 2373 | 97 | |
| GenBank mRNA | BC046514.1 | - | 5 - 6 | 769 | 97 | |
| BC057197.1 | - | 7 - 8 | 1082 | 97 | ||
| BC078445.1 | - | 8 - 9 | 1412 | 97 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
smoothened signaling pathway
cell death
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
nucleotide binding
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
bHLH transcription factor binding
protein phosphorylation
smoothened signaling pathway
cell death
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
nucleotide binding
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
bHLH transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123486 | NM_001081292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104562 | BC078445.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104562 | BC046514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104562 | BC057197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104562 | NM_001081292.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | AK148483.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | BC021452.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | D50494.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | NM_001110826.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | NM_007841.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808686 | NM_181324.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76531 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779351 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78956 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140166 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104562 | BC078445.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104562 | BC046514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104562 | BC057197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104562 | NM_001081292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123486 | NM_001081292.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140166 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01489080 | - | - |
| Whole Mouse Genome G4122A | A_52_P301591 | BC078445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301591 | BC046514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301591 | BC057197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301591 | NM_001081292.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377667 | BC078445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377667 | AK053830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377667 | NM_001081292.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140166 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626839 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05936611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05936615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05936630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06593369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06457749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06443400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06343250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626841 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561330 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457456_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440500_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436373_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561323 | - | - |
| Mouse Genome 430 2.0 Array | 1457456_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436373_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458651_at | - | - |
| Mouse Genome 430 2.0 Array | 1458651_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440500_at | - | - |
| Mouse Genome 430 2.0 Array | 1440500_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457456_at | - | - |
| Mouse Genome 430 2.0 Array | 1436373_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269881_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269881_PM_at | - | - |
| Mouse Expression Set 430 | 1440923_PM_at | - | - |
| Mouse Expression Set 430 | 1436373_PM_at | - | - |
| Mouse Expression Set 430 | 1457456_PM_at | - | - |
| Mouse Expression Set 430 | 1457456_at | - | - |
| Mouse Expression Set 430 | 1440500_at | - | - |
| Mouse Expression Set 430 | 1436373_at | - | - |
| Mouse Expression Set 430 | 1440500_PM_at | - | - |
| Mouse Expression Set 430 | 1440923_at | - | - |
| Murine Genome U74v2 Set | 168942_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168942_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440500_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436373_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440500_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457456_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436373_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457456_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360168 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103940292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613415 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613415 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228169 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103940292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103940292 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228169 | - | - |

Map3k10
Genomic Map