Assay Details
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164147.1 | NP_001157619.1 | 2 - 3 | 462 | 71 | |
NM_001164148.1 | NP_001157620.1 | 2 - 3 | 462 | 71 | ||
NM_001164149.1 | NP_001157621.1 | 2 - 3 | 462 | 71 | ||
NM_001164150.1 | NP_001157622.1 | 2 - 3 | 462 | 71 | ||
NM_001164151.1 | NP_001157623.1 | 2 - 3 | 462 | 71 | ||
NM_001164152.1 | NP_001157624.1 | 2 - 3 | 462 | 71 | ||
NM_001164153.1 | NP_001157625.1 | 2 - 3 | 462 | 71 | ||
NM_011548.4 | NP_035678.3 | 2 - 3 | 462 | 71 | ||
GenBank mRNA | AF352579.1 | - | 2 - 3 | 157 | 71 | |
AK017617.1 | - | 2 - 3 | 430 | 71 | ||
AK034777.1 | - | 2 - 3 | 445 | 71 | ||
AK038738.1 | - | 2 - 3 | 449 | 71 | ||
AK156265.1 | - | 2 - 3 | 445 | 71 | ||
AK161009.1 | - | 2 - 3 | 447 | 71 | ||
AK170674.1 | - | 2 - 3 | 456 | 71 | ||
BC006860.1 | - | 2 - 3 | 416 | 2631291 | 71 | |
BC018260.1 | - | 1 - 2 | 87 | 4187386 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
