Assay Details
| Gene Symbol: Celf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175235.3 | NP_780444.2 | 10 - 11 | 1294 | 62 | |
| GenBank mRNA | AK018246.1 | - | 9 - 10 | 1082 | 62 | |
| AK034216.1 | - | 9 - 10 | 1090 | 62 | ||
| BC052406.1 | - | 10 - 11 | 1294 | 5716119 | 62 | |
| BC057083.1 | - | 9 - 10 | 1076 | 5707546 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Celf6
Genomic Map