Assay Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102455.1 | NP_001095925.1 | 12 - 13 | 1771 | 124 | |
NM_001102456.1 | NP_001095926.1 | 11 - 12 | 1603 | 124 | ||
NM_009691.2 | NP_033821.1 | 11 - 12 | 1603 | 124 | ||
GenBank mRNA | AK149977.1 | - | 12 - 13 | 1772 | 124 | |
AK150020.1 | - | 12 - 13 | 1772 | 124 | ||
AK150478.1 | - | 4 - 5 | 503 | 124 | ||
AK151708.1 | - | 4 - 5 | 503 | 124 | ||
AK152787.1 | - | 4 - 5 | 503 | 124 | ||
AK153087.1 | - | 4 - 5 | 503 | 124 | ||
AK164012.1 | - | 12 - 13 | 1789 | 124 | ||
BC051999.1 | - | 11 - 12 | 1581 | 5716148 | 124 | |
BC052396.1 | - | 11 - 12 | 1575 | 5719918 | 124 | |
M97216.1 | - | 7 - 8 | 963 | 124 | ||
U15571.1 | - | 12 - 13 | 1747 | 124 | ||
Z22592.1 | - | 11 - 12 | 1603 | 124 | ||
Z27070.1 | - | 11 - 12 | 1601 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
