Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011734.3 | NP_035864.2 | 6 - 7 | 920 | 76 | |
| GenBank mRNA | AF156856.1 | - | 7 - 8 | 1122 | 76 | |
| AK028598.1 | - | 6 - 7 | 964 | 76 | ||
| AK028656.1 | - | 6 - 7 | 920 | 76 | ||
| AK033980.1 | - | 168 | 76 | |||
| AK035715.1 | - | 4 - 5 | 654 | 76 | ||
| AK043392.1 | - | 6 - 7 | 941 | 76 | ||
| AK158634.1 | - | 5 - 6 | 858 | 76 | ||
| AK164852.1 | - | 6 - 7 | 927 | 76 | ||
| AK165103.1 | - | 6 - 7 | 947 | 76 | ||
| AK167103.1 | - | 6 - 7 | 936 | 76 | ||
| BC007136.1 | - | 6 - 7 | 938 | 3592286 | 76 | |
| U40408.1 | - | 6 - 7 | 926 | 76 | ||
| U61183.1 | - | 6 - 7 | 919 | 76 | ||
| X98625.1 | - | 6 - 7 | 919 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siae
Genomic Map