Assay Details
| Gene Symbol: Cyp2t4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100184.1 | NP_001093654.1 | 4 - 5 | 524 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2t4| Gene Symbol: Cyp2t4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100184.1 | NP_001093654.1 | 4 - 5 | 524 | 102 | |
Set Membership: |
|