Assay Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029377.2 | NP_083653.1 | 1 - 2 | 207 | 63 | |
| GenBank mRNA | AK007858.1 | - | 1 - 2 | 208 | 63 | |
| AY428767.1 | - | 1 - 2 | 57 | 63 | ||
| BY707863.1 | - | 1 - 2 | 207 | 63 | ||
More Information
Set Membership: |
|

Lin37
Genomic Map