Assay Details
| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021307.2 | NP_067282.2 | 4 - 5 | 386 | 59 | |
| GenBank mRNA | AF167319.1 | - | 3 - 4 | 366 | 59 | |
| BC120886.2 | - | 4 - 5 | 382 | 40060889 | 59 | |
| CN664691.1 | - | 4 - 5 | 447 | 59 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101939 | NM_021307.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101939 | AF167319.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76403 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166127 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113416 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101939 | NM_021307.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101939 | AF167319.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P409729 | AF167319.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25003552 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01484964 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113416 | - | - |
| Whole Mouse Genome G4122A | A_51_P409729 | AF167319.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100360 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113416 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166127 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05991504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06604530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05991514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05991515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05991510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06604530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05926852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06647734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06273737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06655495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05991513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06282643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06230108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06273736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06647735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06230105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06587162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550842 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421458_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550840 | - | - |
| Mouse Genome 430 2.0 Array | 1421458_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421458_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57745_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57745_PM_at | - | - |
| Mouse Expression Set 430 | 1421458_PM_at | - | - |
| Mouse Expression Set 430 | 1421458_at | - | - |
| Murine Genome U74v2 Set | 133153_PM_at | - | - |
| Murine Genome U74v2 Set | 133153_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421458_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421458_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421458_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421458_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 666673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221408 | - | - |
| mouse-6 v1.1 (Illumina) | scl32974.4.1_269-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32974.4.1_269-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221408 | - | - |

Zfp112
Genomic Map