Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033534.1 | NP_001028706.1 | 4 - 5 | 685 | 75 | |
| GenBank mRNA | AK086930.1 | - | 4 - 5 | 646 | 75 | |
| BC139816.1 | - | 657 | 40140447 | 75 | ||
More Information
Set Membership: |
|

Layn
Genomic Map