Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033534.1 | NP_001028706.1 | 5 - 6 | 727 | 119 | |
| GenBank mRNA | BC139816.1 | - | 699 | 40140447 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051834 | BC139816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051834 | AK086930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051834 | NM_001033534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051834 | AK165695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054536 | BC139816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054536 | NM_001033534.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510496 | AF112187.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510496 | AK143995.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510496 | BC021338.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510496 | NM_011326.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510496 | XM_001478438.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411460 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4134 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117323 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144706 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054536 | BC139816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054536 | NM_001033534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051834 | BC139816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051834 | AK086930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051834 | NM_001033534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051834 | AK165695.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144706 | - | - |
| Whole Mouse Genome G4122A | A_51_P236836 | BC139816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236836 | AK086930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236836 | NM_001033534.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117323 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07994761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07759902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08083927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07994762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26507273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07555014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07803746 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10593453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593450 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444165_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442608_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593449 | - | - |
| Mouse Genome 430 2.0 Array | 1444165_at | - | - |
| Mouse Genome 430 2.0 Array | 1442608_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442608_at | - | - |
| Mouse Genome 430 2.0 Array | 1444165_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244864_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244864_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900019065.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019065.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019065.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019066.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019066.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019066.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019068.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019068.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019068.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019063.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019069.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019069.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019069.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019067.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019067.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019067.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019064.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019064.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019064.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019072.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019072.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019072.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019073.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019073.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019073.mm.1 | TC0900002283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019070.mm.1 | NM_001033534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019070.mm.1 | NM_001033534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019070.mm.1 | TC0900002283.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444165_at | - | - |
| Mouse Expression Set 430 | 1444165_PM_at | - | - |
| Mouse Expression Set 430 | 1442608_PM_at | - | - |
| Mouse Expression Set 430 | 1442608_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442608_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444165_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442608_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444165_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 622545 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257873 | - | - |

Layn
Genomic Map