Assay Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145614.4 | NP_663589.3 | 1 - 2 | 586 | 77 | |
| GenBank mRNA | AK032124.1 | - | 1 - 2 | 587 | 77 | |
| AK043771.1 | - | 1 - 2 | 274 | 77 | ||
| AK166090.1 | - | 1 - 2 | 355 | 77 | ||
| BC026680.1 | - | 1 - 2 | 334 | 5059786 | 77 | |
| BC031495.1 | - | 1 - 2 | 307 | 2651095 | 77 | |
| BC069862.1 | - | 1 - 2 | 329 | 5388446 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlat
Genomic Map