Assay Details
| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037143.2 | NP_001032220.1 | 4 - 5 | 495 | 62 | |
| GenBank mRNA | AK009589.1 | - | 4 - 5 | 500 | 62 | |
| AK009604.1 | - | 4 - 5 | 497 | 62 | ||
| AK009778.1 | - | 4 - 5 | 495 | 62 | ||
| AK009924.1 | - | 4 - 5 | 497 | 62 | ||
| BC139039.1 | - | 4 - 5 | 561 | 8862061 | 62 | |
| BC145686.1 | - | 4 - 5 | 561 | 40130989 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211986 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165002 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259689 | AK009924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259689 | BC139039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259689 | BC145686.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259689 | AK009778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P259689 | NM_001037143.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25005457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165002 | - | - |
| Whole Mouse Genome G4122A | A_51_P259689 | AK009924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259689 | BC139039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259689 | BC145686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259689 | AK009778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259689 | NM_001037143.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | AK009924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | BC139039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | BC145686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | AK009778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259694 | NM_001037143.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109583 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05951119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05951125 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05951120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05951122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25005457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05914398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06367119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05914400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25005453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05951120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560864 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429835_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560862 | - | - |
| Mouse Genome 430 2.0 Array | 1429835_at | - | - |
| Mouse Genome 430 2.0 Array | 1429835_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 641361_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 641361_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022475.mm.1 | NM_001037143.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022475.mm.1 | NM_001037143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022475.mm.1 | TC0700002606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022474.mm.1 | NM_001037143.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022474.mm.1 | NM_001037143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022474.mm.1 | TC0700002606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022472.mm.1 | NM_001037143.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022472.mm.1 | NM_001037143.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022472.mm.1 | TC0700002606.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429835_PM_at | - | - |
| Mouse Expression Set 430 | 1429835_at | - | - |
| Murine Genome U74v2 Set | 168004_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168004_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429835_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429835_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429835_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429835_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218348 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245238 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2878161 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878161 | - | - |

Pinlyp
Genomic Map