Assay Details
| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045518.1 | NP_001038983.1 | 1 - 2 | 463 | 133 | |
| GenBank mRNA | AK049628.1 | - | 2 - 3 | 555 | 133 | |
| AK145108.1 | - | 2 - 3 | 547 | 133 | ||
| BC120577.1 | - | 1 - 2 | 463 | 40129500 | 133 | |
| BC120579.1 | - | 1 - 2 | 463 | 40129502 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam83b
Genomic Map