Assay Details
| Gene Symbol: Olfr893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146336.2 | NP_666448.2 | 1 | 691 | 124 | |
| GenBank mRNA | BC130242.1 | - | 1 | 692 | 40142233 | 124 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967578 | NM_146336.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967578 | BC130242.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187371 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164950 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967578 | NM_146336.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967578 | BC130242.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174505 | NM_146336.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174505 | BC130242.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433978 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164950 | - | - |
| Whole Mouse Genome G4122A | A_51_P174505 | NM_146336.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174505 | BC130242.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115104 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584383 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 606690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1500358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237379 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237379 | - | - |
| mouse-6 v1.1 (Illumina) | scl7577.1.1_95-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl7577.1.1_95-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237379 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237379 | - | - |

Olfr893
Genomic Map