Assay Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031141.1 | - | 10 - 11 | 1117 | 73 | |
| AK158516.1 | - | 8 - 9 | 932 | 73 | ||
| AK161676.1 | - | 9 - 10 | 1060 | 73 | ||
| BC017156.1 | - | 10 - 11 | 1238 | 4911201 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp3
Genomic Map