Assay Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031141.1 | - | 1 - 2 | 199 | 127 | |
| AK161676.1 | - | 1 - 2 | 256 | 127 | ||
| BC017156.1 | - | 1 - 2 | 320 | 4911201 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp3
Genomic Map