Assay Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031141.1 | - | 4 - 5 | 478 | 69 | |
| AK158516.1 | - | 2 - 3 | 306 | 69 | ||
| AK161676.1 | - | 4 - 5 | 535 | 69 | ||
| BC017156.1 | - | 4 - 5 | 599 | 4911201 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp3
Genomic Map