Assay Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286544.1 | NP_001273473.1 | 8 - 9 | 2400 | 116 | |
| NM_001286545.1 | NP_001273474.1 | 8 - 9 | 2400 | 116 | ||
| GenBank mRNA | AK134675.1 | - | 8 - 9 | 2297 | 116 | |
| BC043049.1 | - | 8 - 9 | 2299 | 5704185 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccpg1
Genomic Map