Assay Details
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010218.1 | - | 4 - 5 | 763 | 131 | |
AK038631.1 | - | 4 - 5 | 811 | 131 | ||
AK156731.1 | - | 4 - 5 | 737 | 131 | ||
AY176058.1 | - | 4 - 5 | 770 | 131 | ||
AY176059.1 | - | 4 - 5 | 801 | 132 | ||
BC021938.1 | - | 4 - 5 | 768 | 5136809 | 131 | |
U19799.1 | - | 4 - 5 | 782 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
