Assay Details
| Gene Symbol: Thsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040426.3 | NP_001035516.2 | 5 - 6 | 1188 | 68 | |
| GenBank mRNA | AK138976.1 | - | 5 - 6 | 1189 | 68 | |
More Information
Set Membership: |
|

Thsd4
Genomic Map