| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1423889_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10585097 |
- |
- |
| Mouse Genome 430 2.0 Array |
1423889_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1423889_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
434402_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
434402_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900004110.mm.1 |
NM_001004191.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004110.mm.1 |
TC0900000586.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900004109.mm.1 |
NM_001004191.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004109.mm.1 |
TC0900000586.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1423889_PM_at |
- |
- |
| Mouse Expression Set 430 |
1423889_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1423889_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1423889_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1423889_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1423889_at |
- |
- |