Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006431.1 | - | 2 - 3 | 188 | 145 | |
| BC172004.1 | - | 1 - 2 | 136 | 9054290 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC172004.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map