Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 8 - 9 | 1274 | 107 | |
| AK128980.1 | - | 8 - 9 | 1350 | 107 | ||
| BC082577.1 | - | 8 - 9 | 1286 | 30648530 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map