Assay Details
| Gene Symbol: Eml6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146016.2 | NP_666128.2 | 3 - 4 | 1023 | 81 | |
| GenBank mRNA | AK049034.1 | - | 3 - 4 | 935 | 81 | |
| AK049977.1 | - | 2 - 3 | 383 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eml6
Genomic Map