| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100022718.mm.1 |
NM_146016.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022718.mm.1 |
NM_146016.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022718.mm.1 |
TC1100002426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022716.mm.1 |
NM_146016.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022716.mm.1 |
NM_146016.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022716.mm.1 |
TC1100002426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022717.mm.1 |
NM_146016.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022717.mm.1 |
NM_146016.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022717.mm.1 |
TC1100002426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022714.mm.1 |
NM_146016.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022714.mm.1 |
NM_146016.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022714.mm.1 |
TC1100002426.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100022715.mm.1 |
NM_146016.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022715.mm.1 |
NM_146016.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100022715.mm.1 |
TC1100002426.mm.1 |
MTA TC |