Assay Details
| Gene Symbol: Sh3pxd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177364.3 | NP_796338.2 | 5 - 6 | 486 | 68 | |
| GenBank mRNA | AB430861.1 | - | 5 - 6 | 408 | 68 | |
| AK089330.1 | - | 5 - 6 | 458 | 68 | ||
| AK090119.1 | - | 5 - 6 | 479 | 68 | ||
| AK154898.1 | - | 5 - 6 | 444 | 68 | ||
| AK163471.1 | - | 5 - 6 | 458 | 68 | ||
| AK169978.1 | - | 5 - 6 | 478 | 68 | ||
| AK171384.1 | - | 5 - 6 | 458 | 68 | ||
| BC115711.1 | - | 5 - 6 | 455 | 68 | ||
| BC115764.1 | - | 5 - 6 | 455 | 68 | ||
| BC141306.1 | - | 5 - 6 | 458 | 9055918 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3pxd2b
Genomic Map