Assay Details
| Gene Symbol: Sh3pxd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177364.3 | NP_796338.2 | 8 - 9 | 750 | 74 | |
| GenBank mRNA | AB430861.1 | - | 8 - 9 | 672 | 74 | |
| AK089330.1 | - | 8 - 9 | 722 | 74 | ||
| AK090119.1 | - | 8 - 9 | 743 | 74 | ||
| AK154898.1 | - | 8 - 9 | 708 | 74 | ||
| AK171384.1 | - | 8 - 9 | 722 | 74 | ||
| BC115711.1 | - | 8 - 9 | 719 | 74 | ||
| BC115764.1 | - | 8 - 9 | 719 | 74 | ||
| BC141306.1 | - | 8 - 9 | 722 | 9055918 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3pxd2b
Genomic Map