Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146155.3 | NP_666267.3 | 4 - 5 | 536 | 62 | |
| GenBank mRNA | AK149347.1 | - | 4 - 5 | 664 | 62 | |
| BC042621.1 | - | 4 - 5 | 522 | 4911638 | 62 | |
| BC060231.1 | - | 3 - 4 | 504 | 6852595 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC042621.1, BC060231.1, NM_146155.3 transcripts.
Set Membership: |
|

Ahdc1
Genomic Map