| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400012621.mm.1 |
NM_146155.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012621.mm.1 |
NM_146155.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012621.mm.1 |
TC0400001600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012622.mm.1 |
NM_146155.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012622.mm.1 |
NM_146155.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012622.mm.1 |
TC0400001600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012623.mm.1 |
NM_146155.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012623.mm.1 |
NM_146155.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012623.mm.1 |
TC0400001600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012625.mm.1 |
NM_146155.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012625.mm.1 |
NM_146155.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012625.mm.1 |
TC0400001600.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012624.mm.1 |
NM_146155.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012624.mm.1 |
NM_146155.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012624.mm.1 |
TC0400001600.mm.1 |
MTA TC |