Assay Details
| Gene Symbol: Trnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081156.2 | NP_001074625.1 | 1 - 2 | 929 | 110 | |
| GenBank mRNA | AY142705.1 | - | 1 - 2 | 343 | 110 | |
| BC049168.1 | - | 1 - 2 | 885 | 6491759 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
multicellular organism development
nervous system development
cerebellar cortex morphogenesis
regulation of cell proliferation
regulation of cell cycle
neural precursor cell proliferation
DNA binding
protein binding
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
multicellular organism development
nervous system development
cerebellar cortex morphogenesis
regulation of cell proliferation
regulation of cell cycle
neural precursor cell proliferation
DNA binding
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317937 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123673 | NM_001081156.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123673 | AW120970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990373 | AY142705.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990373 | BC049168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990373 | NM_001081156.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | AK004572.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | AK041044.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | AK045181.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | AK049829.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | AK160611.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | BC021899.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324309 | NM_028761.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141574 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157409 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990373 | AY142705.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990373 | BC049168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990373 | NM_001081156.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123673 | NM_001081156.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123673 | AW120970.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00993242 | - | - |
| Whole Mouse Genome G4122A | A_52_P391624 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822110 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822103 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822106 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23317937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23317918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03642786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03481619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03744385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03640231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03707955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03640234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03988089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04060745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03707956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03568331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03754497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04079233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03667987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03408933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03915054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03409112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03707954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03754495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03439765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04079227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03533846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04037971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23317948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517054 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440696_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453008_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517053 | - | - |
| Mouse Genome 430 2.0 Array | 1453008_at | - | - |
| Mouse Genome 430 2.0 Array | 1453008_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440696_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440696_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69539_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69539_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400030352.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030352.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030352.mm.1 | TC0400003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030354.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030354.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030354.mm.1 | TC0400003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030355.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030355.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030355.mm.1 | TC0400003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030350.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030350.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030350.mm.1 | TC0400003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030351.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030351.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030351.mm.1 | TC0400003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030353.mm.1 | NM_001081156.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030353.mm.1 | NM_001081156.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030353.mm.1 | TC0400003702.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440696_at | - | - |
| Mouse Expression Set 430 | 1453008_at | - | - |
| Mouse Expression Set 430 | 1440696_PM_at | - | - |
| Mouse Expression Set 430 | 1453008_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453008_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440696_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440696_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453008_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 316843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253600 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3119914 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3119914 | - | - |

Trnp1
Genomic Map