Assay Details
| Gene Symbol: Kdm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133872.2 | NP_598633.2 | 16 - 17 | 2241 | 84 | |
| GenBank mRNA | AK129170.1 | - | 16 - 17 | 2181 | 84 | |
| BC019417.1 | - | 2 - 3 | 187 | 84 | ||
| BC059885.1 | - | 16 - 17 | 2228 | 6815975 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm1a
Genomic Map