Assay Details
| Gene Symbol: Kdm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133872.2 | NP_598633.2 | 6 - 7 | 1067 | 65 | |
| GenBank mRNA | BC059885.1 | - | 6 - 7 | 1054 | 6815975 | 65 |
More Information
Set Membership: |
|

Kdm1a
Genomic Map