Assay Details
| Gene Symbol: Mad2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019499.4 | NP_062372.2 | 2 - 3 | 286 | 89 | |
| GenBank mRNA | AK010401.1 | - | 2 - 3 | 286 | 88 | |
| AK012590.1 | - | 2 - 3 | 321 | 89 | ||
| AK082934.1 | - | 2 - 3 | 287 | 89 | ||
| AK160286.1 | - | 2 - 3 | 281 | 89 | ||
| AK166065.1 | - | 1 - 2 | 688 | 89 | ||
| BC089012.1 | - | 2 - 3 | 252 | 30539081 | 89 | |
| U83902.1 | - | 2 - 3 | 275 | 89 | ||
More Information
Set Membership: |
|

Mad2l1
Genomic Map