Assay Details
| Gene Symbol: Lrig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008377.2 | NP_032403.2 | 16 - 17 | 3082 | 56 | |
| GenBank mRNA | AK133771.1 | - | 3 - 4 | 650 | 56 | |
| BC141300.1 | - | 16 - 17 | 2874 | 9055912 | 56 | |
| D78572.1 | - | 16 - 17 | 3075 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrig1
Genomic Map