Assay Details
Gene Symbol: Trh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009426.3 | NP_033452.2 | 2 - 3 | 317 | 95 | |
GenBank mRNA | AK010666.1 | - | 2 - 3 | 316 | 95 | |
BC053493.1 | - | 2 - 3 | 303 | 30010395 | 95 | |
CD562178.1 | - | 2 - 3 | 306 | 95 | ||
CF951860.1 | - | 2 - 3 | 305 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
