Assay Details
| Gene Symbol: Lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144799.2 | NP_659048.1 | 4 - 5 | 893 | 60 | |
| GenBank mRNA | AK159946.1 | - | 4 - 5 | 844 | 60 | |
| BC019124.1 | - | 4 - 5 | 844 | 5007525 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmcd1
Genomic Map