Assay Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009662.2 | NP_033792.1 | 11 - 12 | 1728 | 82 | |
| GenBank mRNA | AA388919.1 | - | 2 - 3 | 404 | 82 | |
| AK136868.1 | - | 11 - 12 | 1697 | 82 | ||
| AK137481.1 | - | 11 - 12 | 1686 | 82 | ||
| AK169938.1 | - | 11 - 12 | 1685 | 82 | ||
| AK170434.1 | - | 11 - 12 | 1685 | 82 | ||
| AK170879.1 | - | 11 - 12 | 1684 | 82 | ||
| AK171191.1 | - | 11 - 12 | 1677 | 82 | ||
| AK171413.1 | - | 11 - 12 | 1685 | 82 | ||
| BC139102.1 | - | 11 - 12 | 1655 | 8862124 | 82 | |
| BC141213.1 | - | 11 - 12 | 1655 | 9055825 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alox5
Genomic Map