Assay Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009662.2 | NP_033792.1 | 1 - 2 | 302 | 67 | |
| GenBank mRNA | AK089344.1 | - | 1 - 2 | 256 | 67 | |
| AK136868.1 | - | 1 - 2 | 270 | 67 | ||
| AK137481.1 | - | 1 - 2 | 260 | 67 | ||
| AK169938.1 | - | 1 - 2 | 259 | 67 | ||
| AK170016.1 | - | 1 - 2 | 159 | 67 | ||
| AK170434.1 | - | 1 - 2 | 259 | 67 | ||
| AK170879.1 | - | 1 - 2 | 258 | 67 | ||
| AK171191.1 | - | 1 - 2 | 251 | 67 | ||
| AK171413.1 | - | 1 - 2 | 259 | 67 | ||
| BC139102.1 | - | 1 - 2 | 229 | 8862124 | 67 | |
| BC141213.1 | - | 1 - 2 | 229 | 9055825 | 67 | |
| L42198.1 | - | 1 - 2 | 148 | 67 | ||
More Information
Set Membership: |
|

Alox5
Genomic Map