Assay Details
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027275.3 | NP_081551.2 | 3 - 4 | 219 | 101 | |
GenBank mRNA | AK032831.1 | - | 3 - 4 | 203 | 101 | |
AK146005.1 | - | 3 - 4 | 208 | 101 | ||
AK172480.1 | - | 3 - 4 | 207 | 101 | ||
BB844000.1 | - | 3 - 4 | 220 | 101 | ||
BC115434.2 | - | 3 - 4 | 215 | 40094203 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
