| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_001243741.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_001243741.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_001243742.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_001243742.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_146170.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
NM_146170.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007702.mm.1 |
TC0600000989.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600007690.mm.1 |
NM_146170.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007690.mm.1 |
NM_146170.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007690.mm.1 |
TC0600000989.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600007689.mm.1 |
NM_146170.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007689.mm.1 |
NM_146170.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600007689.mm.1 |
TC0600000989.mm.1 |
MTA TC |