Assay Details
| Gene Symbol: Copg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017477.2 | NP_059505.1 | 4 - 5 | 342 | 88 | |
| NM_201244.1 | NP_957696.1 | 4 - 5 | 310 | 88 | ||
| GenBank mRNA | AF187079.1 | - | 4 - 5 | 341 | 88 | |
| AK049417.1 | - | 4 - 5 | 293 | 88 | ||
| AK077461.1 | - | 4 - 5 | 289 | 88 | ||
| AK080451.1 | - | 4 - 5 | 314 | 88 | ||
| AK147895.1 | - | 4 - 5 | 355 | 88 | ||
| AK151284.1 | - | 4 - 5 | 347 | 88 | ||
| AK151816.1 | - | 4 - 5 | 347 | 88 | ||
| AK153142.1 | - | 4 - 5 | 347 | 88 | ||
| AK164280.1 | - | 4 - 5 | 312 | 88 | ||
| AK169256.1 | - | 4 - 5 | 387 | 88 | ||
| AK171950.1 | - | 4 - 5 | 321 | 88 | ||
| AK172519.1 | - | 4 - 5 | 358 | 88 | ||
| BC024686.1 | - | 4 - 5 | 313 | 3992144 | 88 | |
| BC024896.1 | - | 4 - 5 | 310 | 4189311 | 88 | |
| BC056168.1 | - | 4 - 5 | 310 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copg1
Genomic Map