Assay Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010774.2 | NP_034904.2 | 4 - 5 | 1331 | 106 | |
| GenBank mRNA | AF072249.1 | - | 4 - 5 | 1348 | 106 | |
| BC024812.1 | - | 4 - 5 | 1313 | 5356016 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbd4
Genomic Map