Assay Details
| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101431.1 | NP_001094901.1 | 1 - 2 | 305 | 75 | |
| GenBank mRNA | AA277646.1 | - | 1 - 2 | 304 | 75 | |
| AA833313.1 | - | 1 - 2 | 207 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkrn2os
Genomic Map