Assay Details
| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101431.1 | NP_001094901.1 | 3 - 4 | 509 | 83 | |
| GenBank mRNA | AA833313.1 | - | 3 - 4 | 411 | 83 | |
| AK011120.1 | - | 2 - 3 | 432 | 83 | ||
| AK028760.1 | - | 2 - 3 | 382 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AB195429.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AF416980.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AF421198.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AF421199.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AF421200.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AK036455.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | AK140349.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | BC051526.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | BC132422.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | NM_001039149.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569277 | NM_178687.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96168 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06036338 | - | - |
| Whole Mouse Genome G4122A | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P512427 | AK011120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P512427 | AK028760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P512427 | NM_001101431.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06036338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24771974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05571902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05267388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05213509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05688688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05370946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05853401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05780341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05086769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05853394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05853403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24771910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547031 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440916_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547030 | - | - |
| Mouse Genome 430 2.0 Array | 1440916_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440916_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70291_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70291_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600022678.mm.1 | NM_001101431.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022678.mm.1 | NM_001101431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022678.mm.1 | TC0600002954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600022677.mm.1 | NM_001101431.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022677.mm.1 | NM_001101431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022677.mm.1 | TC0600002954.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440916_PM_at | - | - |
| Mouse Expression Set 430 | 1440916_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440916_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440916_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 660865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216056 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216056 | - | - |

Mkrn2os
Genomic Map