Assay Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013586.4 | NP_038614.2 | 3 - 4 | 571 | 60 | |
| GenBank mRNA | AF053368.1 | - | 3 - 4 | 602 | 60 | |
| AK030548.1 | - | 3 - 4 | 572 | 60 | ||
| AK145189.1 | - | 3 - 4 | 591 | 60 | ||
| BC011298.1 | - | 3 - 4 | 565 | 3488978 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Loxl3
Genomic Map