Assay Details
| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026744.3 | NP_081020.1 | 2 - 3 | 211 | 67 | |
| GenBank mRNA | AK003549.1 | - | 2 - 3 | 212 | 67 | |
| BC022162.1 | - | 2 - 3 | 236 | 5148541 | 67 | |
| BC065995.1 | - | 1434 | 5688315 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591484 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591425 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162616 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162616 | - | - |
| Whole Mouse Genome G4122A | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | NM_026744.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | AK003549.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | BC022162.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385635 | BC065995.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124913 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122328 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162616 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05593113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05368195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05368194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591484 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05078238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05078235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05078226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05037189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513824 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539333 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539321 | - | - |
| Mouse Genome 430 2.0 Array | 1451178_at | - | - |
| Mouse Genome 430 2.0 Array | 1451178_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68499_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68499_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006947.mm.1 | NM_026744.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006947.mm.1 | NM_026744.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006947.mm.1 | TC0600000921.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451178_at | - | - |
| Mouse Expression Set 430 | 1451178_PM_at | - | - |
| Murine Genome U74v2 Set | 99153_PM_at | - | - |
| Murine Genome U74v2 Set | 99153_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451178_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451178_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 501879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6770070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2767187 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2767187 | - | - |
| mouse-6 v1.1 (Illumina) | scl29852.1_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29852.1_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2767187 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2767187 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29852.1_47-S | - | - |

Mrpl53
Genomic Map