Assay Details
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077705.2 | NP_001071173.1 | 6 - 7 | 947 | 109 | |
NM_013545.3 | NP_038573.2 | 6 - 7 | 945 | 109 | ||
GenBank mRNA | AK132509.1 | - | 6 - 7 | 864 | 109 | |
AK150509.1 | - | 6 - 7 | 852 | 109 | ||
AK151077.1 | - | 6 - 7 | 866 | 109 | ||
AK151623.1 | - | 6 - 7 | 869 | 109 | ||
AK159686.1 | - | 6 - 7 | 904 | 109 | ||
AK159833.1 | - | 6 - 7 | 897 | 109 | ||
AK169804.1 | - | 6 - 7 | 947 | 109 | ||
BC012660.1 | - | 6 - 7 | 890 | 4010462 | 109 | |
M68902.1 | - | 6 - 7 | 897 | 109 | ||
M90389.1 | - | 6 - 7 | 847 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
