Assay Details
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077705.2 | NP_001071173.1 | 7 - 8 | 1048 | 61 | |
NM_013545.3 | NP_038573.2 | 7 - 8 | 1046 | 61 | ||
GenBank mRNA | AK132509.1 | - | 7 - 8 | 965 | 61 | |
AK150509.1 | - | 7 - 8 | 953 | 61 | ||
AK151077.1 | - | 7 - 8 | 967 | 61 | ||
AK151623.1 | - | 7 - 8 | 970 | 61 | ||
AK159686.1 | - | 7 - 8 | 1005 | 61 | ||
AK159833.1 | - | 7 - 8 | 998 | 61 | ||
AK167923.1 | - | 7 - 8 | 1000 | 61 | ||
AK169804.1 | - | 7 - 8 | 1048 | 61 | ||
BC012660.1 | - | 7 - 8 | 991 | 4010462 | 61 | |
M68902.1 | - | 7 - 8 | 998 | 61 | ||
M90389.1 | - | 7 - 8 | 948 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
